(±)-Epoxystyrene ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98+%
CAS No.: 96-09-3
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: 2°C to 8°C
Flash Point: 74°(165°F)
Refractive Index: 1.5350
Melting Point: -37°
Boiling Point: 193-196°
Beilstein Reference: 108582
UN No.: 2810
MDL No.: MFCD00005121
Formula:
C₈H₈O Boiling Pt: 193…196 °C Melting Pt: –37 °C Density: 1,053 Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00005121 CAS Number: 96-09-3 EINECS: 202-476-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...