Dibenzyl phosphonate 90+%

Supplier: Thermo Scientific
Warning

Synonyms: Dibenzyl phosphite

L09858.14 L09858.06 L09858.22
L09858.14EA 72.1 EUR
L09858.14 L09858.06 L09858.22
Dibenzyl phosphonate 90+%
Dibenzyl phosphonate

Purity: 90+% CAS No.: 17176-77-1 Molecular Formula: C14H15O3P Formula Weight: 262.25 Flash Point: 110°(230°F) Refractive Index: 1.5540 Odour: Odourless Boiling Point: 110-120°/0.1mm Beilstein Reference: 1982794 Merck Reference: 14,3014 MDL No.: MFCD00004774

Formula: C₁₄H₁₅O₃P
MW: 262,25 g/mol
Boiling Pt: 110…120 °C (0,1 mmHg)
Density: 1,187
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004774
CAS Number: 17176-77-1
EINECS: 241-226-1
Merck Index: 14,3014

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR