(±)-Epoxystyrene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Styrene oxide
MQ200
8.03202.0250
8.03202.0025
8.03202.0250EA
70.8
EUR
8.03202.0250
8.03202.0025
(±)-Epoxystyrene, Sigma-Aldrich®
(±)-Epoxystyrene
Formula:
C₈H₈O MW: 120.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005121 CAS Number: 96-09-3 EINECS: 202-476-7 UN: 2810 ADR: 6.1 III |
Specification Test Results
Solubility | 3 g/l (20 °C) |
Melting Point | -37 °C |
Molar Mass | 120.15 g/mol |
Boiling Point | 193 - 195 °C (1013 hPa) |
Vapor Pressure | 0.4 hPa (20 °C) |
Flash Point | 74 °C |
Refractive Index | 1.5338 (20 °C, 589 nm) |
Explosion Limit | 1.1 - 22 %(V) |
Density | 1.05 g/cm3 (20 °C) |
Ignition Point | 498 °C |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...