Sodium-3-nitrobenzenesulphonate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.06548.0100 8.06548.1000
8.06548.0100EA 27.8 EUR
8.06548.0100 8.06548.1000
Sodium-3-nitrobenzenesulphonate, Sigma-Aldrich®
Sodium 3-nitrobenzenesulphonate
Formula: C₆H₄NNaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00007490
CAS Number: 127-68-4
EINECS: 204-857-3

Order Now

Specification Test Results

Solubility 200 g/l (20 °C)
Melting Point 350 °C (decomposition)
Molar Mass 225.16 g/mol
Bulk Density 450 kg/m3
pH 8 (50 g/l, H2O, 23 °C)
Ignition Point 355 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR