Polyphosphoric acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.07471.0100
8.07471.0500
8.07471.2500
8.07471.9025
8.07471.0100EA
49.2
EUR
8.07471.0100
8.07471.0500
8.07471.2500
8.07471.9025
Polyphosphoric acid, Sigma-Aldrich®
Polyphosphoric acid
Formula:
H(PO₃H)nOH Storage Temperature: Ambient |
MDL Number:
MFCD00084480 CAS Number: 8017-16-1 EINECS: 232-417-0 UN: 3264 ADR: 8 II |
Specification Test Results
Solubility | (20 °C) soluble,Hydrolysis |
Melting Point | -20 °C |
Boiling Point | 530 °C (1013 hPa) |
Vapor Pressure | 2 hPa (20 °C) |
Density | 2.06 g/cm3 (20 °C) |
pH | (H2O, 20 °C) acidic,Hydrolysis |
Synonyms | PPA |
Hill Formula | HO[P(OH)(O)O](n)H |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...