Thallium(I) sulphate ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Thallic sulphate, Dithallium sulphate

40342.18 40342.09 40342.30 40342.A1
40342.18EA 540 EUR
40342.18 40342.09 40342.30 40342.A1
Thallium(I) sulphate ≥99.5% (metals basis)
Dithallium sulphate

Purity: 99.5% min CAS No.: 7446-18-6 Molecular Formula: Tl2SO4 Formula Weight: 504.80 Physical Form: Crystalline Density: 6.77 Melting Point: 632° Boiling Point: dec. Solubility: Solubility in water increases with temperature Merck Reference: 14,9269 UN No.: 1707 MDL No.: MFCD00011278

Formula: O₄STl₂
Boiling Pt: Dec.
Melting Pt: 632 °C
Density: 6,77
Storage Temperature: Ambient
MDL Number: MFCD00011278
CAS Number: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1,II
Merck Index: 14,9269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR