Germanium(IV) chloride 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
197150250
197151000
197150050
ACRO197150250EA
285
EUR
ACRO197150250
ACRO197151000
ACRO197150050
Germanium(IV) chloride 99.99% (trace metals basis)
Germanium (IV) chloride
Formula:
Cl₄Ge Boiling Pt: 82 °C (1013 hPa) Melting Pt: –49,5 °C Density: 1,84 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011029 CAS Number: 10038-98-9 EINECS: 233-116-7 UN: 2922 ADR: 8,I Merck Index: 12,04418 |
Specification Test Results
Appearance | Clear colorless Liquid |
Total trace metal impurities | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...