Antimony(III) chloride ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Antimony trichloride

A11944.0E A11944.22 A11944.36
A11944.0EEA 411 EUR
A11944.0E A11944.22 A11944.36
Antimony(III) chloride ≥99%
Antimony (III) chloride

Purity: 99+% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Odour: Pungent Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 223.5 °C
Melting Pt: 73.4 °C
Density: 3.14
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8 II
Merck Index: 13,00713

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR