Tellurium tetrabromide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific
Danger

Synonyms: Tellurium(IV) bromide

ampouled under argon

43439.09 43439.18
43439.09EA 199 EUR
43439.09 43439.18
Tellurium tetrabromide, ultra dry ≥99.999% (metals basis)
Tellurium tetrabromide

Purity: 99.999% CAS No.: 10031-27-3 Molecular Formula: TeBr4 Formula Weight: 447.27 Physical Form: Powder, Ampouled under argon Density: 4.31 Melting Point: 380° Boiling Point: 420° dec. Merck Reference: 14,9125 UN No.: 2923 MDL No.: MFCD00049575

Formula: Br₄Te
MW: 447,22 g/mol
Boiling Pt: 420 °C (dec.)
Melting Pt: 380 °C
Density: 4,31
Storage Temperature: Ambient
MDL Number: MFCD00049575
CAS Number: 10031-27-3
EINECS: 233-090-7
UN: 2923
ADR: 8,II
Merck Index: 14,9125

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR