Selenium tetrachloride ≥99.5% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Selenium(IV) chloride
13096.14
13096.22
13096.14EA
155
EUR
13096.14
13096.22
Selenium tetrachloride ≥99.5% (metals basis)
Selenium tetrachloride
Purity: 99.5%
CAS No.: 10026-03-6
Molecular Formula: SeCl4
Formula Weight: 220.77
Physical Form: Lump
Density: 2.600
Refractive Index: 1.807
Sensitivity: Moisture Sensitive
Odour: Pungent
Melting Point: subl.
Solubility: Decomposes in water and moist air. Insoluble in liquid bromine. Sublimes when heated
Merck Reference: 14,8438
UN No.: 2923
MDL No.: MFCD00011225
Formula:
Cl₄Se MW: 220,77 g/mol Melting Pt: (subl.) Density: 2,600 Storage Temperature: Ambient |
MDL Number:
MFCD00011225 CAS Number: 10026-03-6 EINECS: 233-053-5 UN: 2923 ADR: 8,II Merck Index: 14,8438 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...