Tellurium dioxide ≥99.9995% (metals basis), Puratronic®
Supplier: Thermo Scientific
|
Warning
|
10882.14
10882.22
10882.36
10882.14EA
161
EUR
10882.14
10882.22
10882.36
Tellurium dioxide ≥99.9995% (metals basis), Puratronic®
Tellurium dioxide
Purity: 99.9995%
CAS No.: 7446-07-3
Molecular Formula: TeO2
Formula Weight: 159.60
Physical Form: Powder
Density: 5.8
Melting Point: 733°
Merck Reference: 14,9123
MDL No.: MFCD00011263
Formula:
O₂Te MW: 159,6 g/mol Boiling Pt: 1245 °C (1013 hPa) Melting Pt: 733 °C Density: 5,8 Storage Temperature: Ambient |
MDL Number:
MFCD00011263 CAS Number: 7446-07-3 EINECS: 231-193-1 Merck Index: 14,9123 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...