Tellurium dioxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

1.12356.0025 1.12356.0010
1.12356.0025EA 112 EUR
1.12356.0025 1.12356.0010
Tellurium dioxide, Sigma-Aldrich®
Tellurium dioxide
Formula: O₂Te
Boiling Pt: 1245 °C (1013 hPa)
Melting Pt: 773 °C
Density: 5,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 733 °C
Molar Mass 159.60 g/mol
Boiling Point 1245 °C
Density 6.02 g/cm3 (20 °C)
Synonyms Tellurium(IV) oxide
Hill Formula O2Te
Chemical Formula TeO2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR