Dimethyl diselenide ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Dimethyldiselenide
68119.03
68119.06
68119.03EA
92.5
EUR
68119.03
68119.06
Dimethyl diselenide ≥99%
Dimethyl diselenide
Purity: 99%
CAS No.: 7101-31-7
Molecular Formula: (CH3)2Se2
Formula Weight: 187.99
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.987
Refractive Index: 1.6388
Sensitivity: Air and Moisture Sensitive
Odour: Unpleasant
Boiling Point: 155-157°
UN No.: 2929
MDL No.: MFCD00008254
Formula:
C₂H₆Se₂ MW: 187.99 g/mol Boiling Pt: 155…157 °C Melting Pt: –82 °C Density: 1.987 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008254 CAS Number: 7101-31-7 EINECS: 230-410-7 UN: 2929 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...