2-Indanol ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Hydroxyindane
L01766.14
L01766.22
L01766.14EA
0
EUR
L01766.14
L01766.22
2-Indanol ≥99%
2-Indanol
Purity: 99%
CAS No.: 4254-29-9
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 109°(228°F)
pH: 7
Melting Point: 67-72°
Boiling Point: 120-123°/12mm
Beilstein Reference: 1862567
MDL No.: MFCD00003800
Formula:
C₉H₁₀O MW: 134,18 g/mol Boiling Pt: 120…123 °C (12 mmHg) Melting Pt: 67…72 °C Density: 1,0528 g/cm³ (80 °C) Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003800 CAS Number: 4254-29-9 EINECS: 224-230-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...