Indane 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2,3-Dihydro-1H-indene, Indan

292480050 292481000 292485000
ACRO292480050EA 44.3 EUR
ACRO292480050 ACRO292481000 ACRO292485000
Indane 95%
Indane
Formula: C₉H₁₀
MW: 118,18 g/mol
Boiling Pt: 176,5 °C (1013 hPa)
Melting Pt: –51 °C
Density: 0,96 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00003795
CAS Number: 496-11-7
EINECS: 207-814-7
UN: 1993
ADR: 3,III
Merck Index: 13,04956

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5380 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR