4-Bromoisoquinoline 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
107090010
107090100
ACRO107090010EA
30.5
EUR
ACRO107090010
ACRO107090100
4-Bromoisoquinoline 98%
4-Bromoisoquinoline
Formula:
C₉H₆BrN MW: 208.06 g/mol Boiling Pt: 280 Melting Pt: 39…43 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006904 CAS Number: 1532-97-4 EINECS: 216-244-8 UN: 2811 |
Specification Test Results
Appearance | White to light beige Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 39 to 43 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...