4-Bromoisoquinoline ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A15951.06
A15951.14
A15951.22
A15951.06EA
63.1
EUR
A15951.06
A15951.14
A15951.22
4-Bromoisoquinoline ≥98%
4-Bromoisoquinoline
Purity: 98%
CAS No.: 1532-97-4
Molecular Formula: C9H6BrN
Formula Weight: 208.06
Flash Point:
Formula:
C₉H₆BrN Boiling Pt: 285 °C Melting Pt: 39…43 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006904 CAS Number: 1532-97-4 EINECS: 216-244-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...