Silver nitrate BP, Ph. Eur., USP, extra pure, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
|
Danger
|
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
1.01510.0050
1.01510.0250
1.01510.1000
1.01510.0050EA
631
EUR
1.01510.0050
1.01510.0250
1.01510.1000
Silver nitrate BP, Ph. Eur., USP, extra pure, SAFC®
Silver nitrate
Formula:
AgNO₃ Boiling Pt: 444 °C (1013 hPa) Melting Pt: 210 °C Density: 4,35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1493 ADR: 5.1,II REACH: 01-2119513705-43 Merck Index: 12,08661 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...