Silver perrhenate ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Silver(I) perrhenate
44480.03
44480.06
44480.03EA
87.8
EUR
44480.03
44480.06
Silver perrhenate ≥99.99% (metals basis)
Silver perrhenate
Purity: 99.99%
CAS No.: 20654-56-2
Molecular Formula: AgO4Re
Formula Weight: 358.10
Physical Form: Powder
Density: 7.05
Appearance: White
Melting Point: 430°
UN No.: 1479
MDL No.: MFCD00014144
Formula:
AgO₄Re Melting Pt: 430 °C Density: 7,05 Storage Temperature: Ambient |
MDL Number:
MFCD00014144 CAS Number: 7784-00-1 EINECS: 232-042-2 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...