Trizinc diarsenide ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
35660.03
35660.06
35660.03EA
0
EUR
35660.03
35660.06
Trizinc diarsenide ≥99.999% (metals basis)
Trizinc diarsenide
Purity: 99.999%
CAS No.: 12006-40-5
Molecular Formula: Zn3As2
Formula Weight: 345.95
Physical Form: 6.35mm & Down Pieces
Density: 5.528
Melting Point: 1015°
UN No.: 1557
MDL No.: MFCD00049629
Formula:
As₂Zn₃ Melting Pt: 1015 °C Density: 5,528 Storage Temperature: Ambient |
MDL Number:
MFCD00049629 CAS Number: 12006-40-5 EINECS: 234-486-2 UN: 1557 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...