Mercury selenide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
22878.04
22878.09
22878.04EA
120
EUR
22878.04
22878.09
Mercury selenide ≥99.9% (metals basis)
Mercury selenide
Purity: 99.9%
CAS No.: 20601-83-6
Molecular Formula: HgSe
Formula Weight: 279.55
Physical Form: -100 Mesh Powder
Density: 8.21
Appearance: Gray
Odour: Odourless
Melting Point: subl.
UN No.: 2025
MDL No.: MFCD00049743
Formula:
HgSe MW: 279.55 g/mol Melting Pt: 998 (subl.) Density: 8.21 Storage Temperature: Ambient |
MDL Number:
MFCD00049743 CAS Number: 20601-83-6 EINECS: 243-910-5 UN: 2025 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...