Rhenium trichloride
Supplier: Thermo Fisher Scientific
|
Warning
|
13230.01
13230.03
13230.01EA
187
EUR
13230.01
13230.03
Rhenium trichloride
Rhenium trichloride
CAS No.: 13569-63-6
Molecular Formula: ReCl3
Formula Weight: 292.56
Physical Form: Powder
Sensitivity: Moisture Sensitive
Odour: Odourless
MDL No.: MFCD00011196
Formula:
Cl₃Re MW: 292.57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011196 CAS Number: 13569-63-6 EINECS: 236-987-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...