Manganese(II) telluride ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Manganese telluride

36389.09 36389.03
36389.09EA 462 EUR
36389.09 36389.03
Manganese(II) telluride ≥99.9% (metals basis)
Manganese (II) telluride

Purity: 99.9% CAS No.: 12032-88-1 Molecular Formula: MnTe + small amount MnTe2 Formula Weight: 182.54 Physical Form: 10mm & Down Lump Appearance: Dark grey MDL No.: MFCD00049487

Formula: MnTe
Melting Pt: 1150 °C
Density: 6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049487
CAS Number: 12032-88-1
EINECS: 234-782-1

Order Now

Specification Test Results

Total Metal Impurities 0,1% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR