1-Naphthaldehyde 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
α-Naphthaldehyde
128091000
128095000
128090025
ACRO128091000EA
99.9
EUR
ACRO128091000
ACRO128095000
ACRO128090025
1-Naphthaldehyde 95%
1-Naphthaldehyde
Formula:
C₁₁H₈O MW: 156.18 g/mol Boiling Pt: 160…161 °C (15 mmHg) Melting Pt: 1…2 °C Density: 1.15 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004003 CAS Number: 66-77-3 EINECS: 200-633-4 UN: 3334 ADR: 9 III |
Specification Test Results
Appearance | Clear yellow to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6500 to 1.6540 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...