2-Chloronaphthalene
Supplier: LGC Standards PROMOCHEM
CILDLM20050.01
CILDLM20050.1
PROOCILDLM20050.01EA
771
EUR
PROOCILDLM20050.01
PROOCILDLM20050.1
2-Chloronaphthalene
2-Chloronaphthalene
Formula:
C₁₀H₇Cl MW: 162.62 g/mol Boiling Pt: 256 Melting Pt: 59.5 Density: 1.2656 g/cm³ (16 °C) |
MDL Number:
MFCD00035731 CAS Number: 91-58-7 EINECS: 202-079-9 Merck Index: 13,02169 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...