1-Iodonaphthalene 97.5%
Supplier: Thermo Fisher Scientific
|
Warning
|
218420100
218420500
ACRO218420100EA
75.8
EUR
ACRO218420100
ACRO218420500
1-Iodonaphthalene 97.5%
1-Iodonaphthalene
Formula:
C₁₀H₇I Boiling Pt: 302…305 °C (760 mmHg) Melting Pt: 7 °C Density: 1,74 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003876 CAS Number: 90-14-2 EINECS: 201-968-9 |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...