1-Iodonaphthalene ≥98%

Supplier: Thermo Fisher Scientific
Warning

B22644.09 B22644.18
B22644.09EA 78.1 EUR
B22644.09 B22644.18
1-Iodonaphthalene ≥98%
1-Iodonaphthalene

Purity: 98% CAS No.: 90-14-2 Molecular Formula: C10H7I Formula Weight: 254.07 Flash Point: 110°(230°F) Refractive Index: 1.7010 Melting Point: 4° Boiling Point: 161-162°/15mm Beilstein Reference: 1906415 MDL No.: MFCD00003876

Formula: C₁₀H₇I
Boiling Pt: 161…162 °C (15 mmHg)
Melting Pt: 4 °C
Density: 1,738
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003876
CAS Number: 90-14-2
EINECS: 201-968-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR