1-Bromo-2-methylnaphthalene 90%, Technical Grade
Supplier: Thermo Fisher Scientific
|
Warning
|
154260250
154261000
ACRO154260250EA
97.9
EUR
ACRO154260250
ACRO154261000
1-Bromo-2-methylnaphthalene 90%, Technical Grade
1-Bromo-2-methylnaphthalene
Formula:
C₁₁H₉Br MW: 221.1 g/mol Boiling Pt: 296 °C (760 mmHg) Density: 1.418 g/cm³ Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003871 CAS Number: 2586-62-1 EINECS: 219-966-1 |
Specification Test Results
Appearance | Clear colorless to yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6450 to 1.6530 (20°C, 589 nm) |
GC | ≥88 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...