2-(Chloromethyl)naphthalene ≥98%
Supplier: Thermo Scientific
|
Danger
|
H50242.03
H50242.03EA
276
EUR
H50242.03
2-(Chloromethyl)naphthalene ≥98%
2-(Chloromethyl)naphthalene
Purity: 98%
CAS No.: 2506-41-4
Molecular Formula: C11H9Cl
Formula Weight: 176.65
Melting Point: 47-50°
UN No.: 3261
MDL No.: MFCD00046488
Formula:
C₁₁H₉Cl MW: 176,65 g/mol Melting Pt: 47…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046488 CAS Number: 2506-41-4 EINECS: 219-713-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...