1-Methylnaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.20809.0005
8.20809.0250
8.20809.1000
8.20809.0005EA
38.2
EUR
8.20809.0005
8.20809.0250
8.20809.1000
1-Methylnaphthalene for synthesis, Sigma-Aldrich®
1-Methylnaphthalene
Formula:
C₁₁H₁₀ Boiling Pt: 244…245 °C (1013 hPa) Melting Pt: –33…–32 °C Density: 1,022 g/cm³ (20 °C) Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004034 CAS Number: 90-12-0 EINECS: 201-966-8 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | 0.026 g/l (25 °C) |
Molar Mass | 142.20 g/mol |
Boiling Point | 244 - 245 °C (1013 mbar) |
Vapor Pressure | 2 hPa (25 °C) |
Flash Point | 82 °C |
Explosion Limit | 0.7 - 6.5 %(V) |
Density | 1.02 g/cm3 (20 °C) |
Ignition Point | 529 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...