2-Bromo-6-chloropyridine 95%

Supplier: Thermo Fisher Scientific
Danger

H26961.03 H26961.MD
H26961.03EA 0 EUR
H26961.03 H26961.MD
2-Bromo-6-chloropyridine 95%
2-Bromo-6-chloropyridine

Purity: 95% CAS No.: 5140-72-7 Molecular Formula: C5H3BrClN Formula Weight: 192.44 MDL No.: MFCD00181262

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00181262
CAS Number: 5140-72-7
EINECS: 225-904-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR