2-Chloropyridine 99%

Supplier: Thermo Fisher Scientific
Danger

110011000 110015000
ACRO110011000EA 34.7 EUR
ACRO110011000 ACRO110015000
2-Chloropyridine 99%
2-Chloropyridine
Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 168…170 °C (760 mmHg)
Melting Pt: –46 °C
Density: 1.209 g/cm³
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00006228
CAS Number: 109-09-1
EINECS: 203-646-3
UN: 2822
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5310 to 1.5330 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.5 %
Visual transition interval from pH 1.2 (red) to 2.3 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR