2-Chloropyridine ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
A14866.22
A14866.36
A14866.22EA
38.6
EUR
A14866.22
A14866.36
2-Chloropyridine ≥99%
2-Chloropyridine
Purity: 99%
CAS No.: 109-09-1
Molecular Formula: C5H4ClN
Formula Weight: 113.55
Flash Point: 65°(149°F)
Refractive Index: 1.5320
Melting Point: -46°
Boiling Point: 167-170°
Beilstein Reference: 105788
UN No.: 2822
MDL No.: MFCD00006228
Formula:
C₅H₄ClN MW: 113.55 g/mol Boiling Pt: 167…170 °C Melting Pt: –46 °C Density: 1.208 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006228 CAS Number: 109-09-1 EINECS: 203-646-3 UN: 2822 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...