3-Chloropyridine ≥99%

Supplier: Thermo Scientific
Danger

B24316.09 B24316.18
B24316.09EA 35.5 EUR
B24316.09 B24316.18
3-Chloropyridine ≥99%
3-Chloropyridine

Purity: 99% CAS No.: 626-60-8 Molecular Formula: C5H4ClN Formula Weight: 113.55 Flash Point: 53°(127°F) Refractive Index: 1.5330 Boiling Point: 147-149° Beilstein Reference: 105744 UN No.: 1993 MDL No.: MFCD00006375

Formula: C₅H₄ClN
MW: 113,55 g/mol
Boiling Pt: 147…149 °C
Melting Pt: –0,5 °C
Density: 1,205
Flash Pt: 53 °C (127 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006375
CAS Number: 626-60-8
EINECS: 210-955-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR