3-Iodopyridine ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23225.14
B23225.06
B23225.14EA
375
EUR
B23225.14
B23225.06
3-Iodopyridine ≥99%
3-Iodopyridine
Purity: 99%
CAS No.: 1120-90-7
Molecular Formula: C5H4IN
Formula Weight: 205.00
Flash Point: 106°(222°F)
Melting Point: 53-56°
Boiling Point: 90-92°/14mm
Beilstein Reference: 106059
MDL No.: MFCD00023553
Formula:
C₅H₄IN MW: 205 g/mol Boiling Pt: 90…92 °C (14 mmHg) Melting Pt: 53…56 °C Flash Pt: 106 °C (222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00023553 CAS Number: 1120-90-7 EINECS: 214-322-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...