2-Aminopyridine 99+%
Supplier: Thermo Fisher Scientific
|
Danger
|
104541000
104545000
104540025
ACRO104541000EA
51.4
EUR
ACRO104541000
ACRO104545000
ACRO104540025
2-Aminopyridine 99+%
2-Aminopyridine
Formula:
C₅H₆N₂ Boiling Pt: 204…210 °C (760 mmHg) Melting Pt: 55…60 °C Density: 1,24 g/cm³ (20 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006312 CAS Number: 504-29-0 EINECS: 207-988-4 UN: 2671 ADR: 6.1,II Merck Index: 13,00472 |
Specification Test Results
Appearance | White to yellow to light beige Powder or crystals or flakes and/or chunks |
Infrared spectrum | Conforms |
Melting point | 55 to 60 °C |
GC | ≥99.0 % |
Moisture | ≤3.0 % (As packaged) |
Viscosity | 0.10 to 0.20 s |
Hydroxyl content | 4.4 to 5.6 % |
Free acid (CH3COOH) | ≤0.04 % |
Potassium (K) | ≤0.01 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.01 % |
Loss on ignition | ≤0.5 % |
Nitrogen compounds (as N) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...