2-Bromo-5-methylpyridine ≥98%
Supplier: Thermo Scientific
|
Danger
|
B25083.06
B25083.14
B25083.22
B25083.06EA
52.1
EUR
B25083.06
B25083.14
B25083.22
2-Bromo-5-methylpyridine ≥98%
2-Bromo-5-methylpyridine
Purity: 98+%
CAS No.: 3510-66-5
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 103°(217°F)
Melting Point: 40-45°
Boiling Point: 95-96°/12mm
Beilstein Reference: 107323
MDL No.: MFCD00209553
Formula:
C₆H₆BrN Boiling Pt: 95…96 °C (12 mmHg) Melting Pt: 40…45 °C Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00209553 CAS Number: 3510-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...