2-Pentylpyridine ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Amylpyridine
B22125.06
B22125.14
B22125.06EA
51.7
EUR
B22125.06
B22125.14
2-Pentylpyridine ≥98%
2-Pentylpyridine
Purity: 98%
CAS No.: 2294-76-0
Molecular Formula: C10H15N
Formula Weight: 149.24
Flash Point: 79°(174°F)
Refractive Index: 1.4890
Odour: Pleasant
Boiling Point: 203-204°
Beilstein Reference: 2772
MDL No.: MFCD00051828
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Boiling Pt: 203…204 °C Density: 0.903 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051828 CAS Number: 2294-76-0 EINECS: 218-937-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...