4-Picoline ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Methylpyridine, γ-Picoline
A15595.0F
A15595.AE
A15595.AP
A15595.0FEA
163
EUR
A15595.0F
A15595.AE
A15595.AP
4-Picoline ≥98%
4-Picoline
Purity: 98%
CAS No.: 108-89-4
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 39°(102°F)
Refractive Index: 1.5050
Odour: Unpleasant
pH: 10-11
Melting Point: 3°
Boiling Point: 144-145°
Beilstein Reference: 104586
Merck Reference: 14,7402
UN No.: 2313
MDL No.: MFCD00006440
Formula:
C₆H₇N MW: 93,13 g/mol Boiling Pt: 144…145 °C Melting Pt: 3 °C Density: 0,957 Flash Pt: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006440 CAS Number: 108-89-4 EINECS: 203-626-4 UN: 2313 ADR: 3,III Merck Index: 14,7402 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...