Thiophene 99.5%, extra pure benzene free
Supplier: Thermo Fisher Scientific
|
Danger
|
342591000
342595000
ACRO342591000EA
47.2
EUR
ACRO342591000
ACRO342595000
Thiophene 99.5%, extra pure benzene free
Thiophene
Formula:
C₄H₄S Boiling Pt: 84,4 °C (1013 hPa) Melting Pt: –38 °C Density: 1,065 g/cm³ Flash Pt: –9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005413 CAS Number: 110-02-1 EINECS: 203-729-4 UN: 2414 ADR: 3,II Merck Index: 12,09490 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5270 to 1.5290 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥99.4 % |
Color scale | ≤40 APHA |
Benzene | ≤0.1 % (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...