2-Methylthiophene ≥98%
Supplier: Thermo Scientific
|
Danger
|
A12736.14
A12736.22
A12736.36
A12736.14EA
40.8
EUR
A12736.14
A12736.22
A12736.36
2-Methylthiophene ≥98%
2-Methylthiophene
Purity: 98%
CAS No.: 554-14-3
Molecular Formula: C5H6S
Formula Weight: 98.17
Flash Point: 7°(44°F)
Refractive Index: 1.5200
Odour: Aromatic
Melting Point: -63°
Boiling Point: 112-114°
Beilstein Reference: 103734
UN No.: 1993
MDL No.: MFCD00005451
Formula:
C₅H₆S MW: 98,17 g/mol Boiling Pt: 112…114 °C Melting Pt: –63 °C Density: 1,017 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005451 CAS Number: 554-14-3 EINECS: 209-063-0 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...