Tridodecylamine (hydrogen ionophore I), Supelco®
Supplier: Merck
Synonyms:
Trilaurylamine, Tridodecylamine
M-Clarity™ quality level = MQ100
95292-100MG
95292-500MG
95292-100MGEA
124
EUR
95292-100MG
95292-500MG
Tridodecylamine (hydrogen ionophore I), Supelco®
Tridodecylamine (Hydrogen ionophore I)
Formula:
C₃₆H₇₅N Boiling Pt: 450 °C (1013 hPa) Melting Pt: 16 °C Density: 0,82 g/cm³ (20 °C) Flash Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008971 CAS Number: 102-87-4 EINECS: 203-063-4 UN: 3082 ADR: 9,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...