4-Nitrophenyl palmitate 98
Supplier: Thermo Fisher Scientific
Synonyms:
4-Nitrophenyl hexadecanoate
L10896.03
L10896.06
L10896.03EA
39.8
EUR
L10896.03
L10896.06
4-Nitrophenyl palmitate 98
4-Nitrophenyl palmitate
Purity: 98+%
CAS No.: 1492-30-4
Molecular Formula: C22H35NO4
Formula Weight: 377.53
Melting Point: 65-66°
Beilstein Reference: 1891754
Formula:
C₂₂H₃₅NO₄ MW: 377.52 g/mol Melting Pt: 65…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047732 CAS Number: 1492-30-4 EINECS: 216-084-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...