(±)-Ethyl-2-bromopropionate 99%

Supplier: Thermo Fisher Scientific
Danger

118141000 118142500 118140010
ACRO118141000EA 49.8 EUR
ACRO118141000 ACRO118142500 ACRO118140010
(±)-Ethyl-2-bromopropionate 99%
(±)-Ethyl 2-bromopropionate
Formula: C₅H₉BrO₂
Boiling Pt: 156…160 °C (760 mmHg)
Density: 1,394 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥98.5 %
Sulfate (SO4) ≤300 ppm
Sodium fluosilicate ≤0.1 % (Na2SiF6)
Sulfite (SO2) ≤50 ppm
Iron (Fe) ≤30 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Pb) ≤30 ppm
Titrable acid ≤0.03 meq/g
Titrable base ≤0.01 meq/g
Insoluble matter ≤0.02 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR