Sesamol-(3,4-(methylenedioxy)phenol) ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Sesamol, 3,4-Methylenedioxyphenol, Benzo[d][1,3]dioxol-5-ol
A10453.09
A10453.18
A10453.30
A10453.09EA
43.8
EUR
A10453.09
A10453.18
A10453.30
Sesamol-(3,4-(methylenedioxy)phenol) ≥98%
Sesamol (3,4-(Methylenedioxy)phenol)
Purity: 98%
CAS No.: 533-31-3
Molecular Formula: C7H6O3
Formula Weight: 138.12
Storage Temperature: 2°C to 8°C
Melting Point: 61-65°
Boiling Point: 113-116°/2mm
Beilstein Reference: 127405
MDL No.: MFCD00005827
Formula:
C₇H₆O₃ Boiling Pt: 113…116 °C (2 mmHg) Melting Pt: 61…65 °C |
MDL Number:
MFCD00005827 CAS Number: 533-31-3 EINECS: 208-561-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...