1-Aminobenzotriazole 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
348641000
348642500
ACRO348641000EA
89.2
EUR
ACRO348641000
ACRO348642500
1-Aminobenzotriazole 98%
1-Aminobenzotriazole
Formula:
C₆H₆N₄ Melting Pt: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132902 CAS Number: 1614-12-6 |
Specification Test Results
Appearance | Yellow to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 81 to 85 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...