(±)-2-Bromopropionyl bromide ≥97%

Supplier: Thermo Fisher Scientific
Danger

B22504.22 B22504.36
B22504.22EA 45 EUR
B22504.22 B22504.36
(±)-2-Bromopropionyl bromide ≥97%
(±)-2-Bromopropionyl bromide

Purity: 97% CAS No.: 563-76-8 Molecular Formula: CH3CHBrCOBr Formula Weight: 215.88 Flash Point: 110°(230°F) Refractive Index: 1.5210 Boiling Point: 48-50°/10mm Beilstein Reference: 1071331 UN No.: 3265 MDL No.: MFCD00000142

Formula: C₃H₄Br₂O
MW: 215.87 g/mol
Boiling Pt: 48…50 °C (10 mmHg)
Density: 2.061
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000142
CAS Number: 563-76-8
EINECS: 209-261-7
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR