(±)-2-Chloropropionyl chloride 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
295161000
295165000
295160250
ACRO295161000EA
114
EUR
ACRO295161000
ACRO295165000
ACRO295160250
(±)-2-Chloropropionyl chloride 95%
(±)-2-Chloropropionyl chloride
Formula:
C₃H₄Cl₂O Boiling Pt: 109…111 °C (760 mmHg) Melting Pt: –71 °C Density: 1,3 g/cm³ Flash Pt: 34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000862 CAS Number: 7623-09-8 EINECS: 231-540-7 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4390 to 1.4410 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...