(±)-Methyl-2-chloropropionate ≥97%
Supplier: Thermo Scientific
|
Warning
|
A18541.0B
A18541.0I
A18541.30
A18541.0BEA
265
EUR
A18541.0B
A18541.0I
A18541.30
(±)-Methyl-2-chloropropionate ≥97%
(±)-Methyl 2-chloropropionate
Purity: 97%
CAS No.: 17639-93-9
Molecular Formula: C4H7ClO2
Formula Weight: 122.55
Flash Point: 38°(100°F)
Refractive Index: 1.4170
Melting Point:
Formula:
C₄H₇ClO₂ MW: 122,55 g/mol Boiling Pt: 132…133 °C Melting Pt: < –10 °C Density: 1,140 Flash Pt: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000863 CAS Number: 17639-93-9 EINECS: 241-624-5 UN: 2933 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...