Potassium thioacetate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Potassium ethanethioate

221300250 221301000 221305000
ACRO221300250EA 103 EUR
ACRO221300250 ACRO221301000 ACRO221305000
Potassium thioacetate 98%
Potassium thioacetate
Formula: C₂H₃KOS
MW: 114,21 g/mol
Melting Pt: 173…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137704
CAS Number: 10387-40-3
EINECS: 233-848-7
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
Solubility (10 % in water) Clear
pH ≤11 (10 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR